Operating Scissors, Curved | Avantor
Operating Scissors, Curved
Operating scissors, sharp/sharp, 11.5 cm (4.5"), Stainless steel, Curved
Catalog # 77662-808
Fournisseur #:  501755
Operating Scissors, Curved
Catalog # 77662-808
Operating Scissors, Curved
Operating scissors, sharp/sharp, 11.5 cm (4.5"), Stainless steel, Curved
Price
$90.03
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ring-handled operating scissors are a pair of curved surgical scissors commonly used in general surgery for cutting and dissecting tissues.
  • Sharp/sharp, Sharp/blunt, or Blunt/blunt tips
  • Stainless steel or premium German steel
  • Autoclavable
The 11.5 cm length is a smaller size that can be used for a variety of procedures, including biological and medical research. These operating scissors are curved giving a better view of the subject. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries.
These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades. They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion.
Operating scissors are designed to withstand the high heat and chemical exposure of the sterilization process.
Spécifications
  • Désignation:
    Operating scissors, sharp/sharp
  • Longueur:
    11.5 cm (4.5")
  • Matière:
    Stainless steel
  • Curvature:
    Curved