Mayo Scissors Supercut
Mayo Scissors Supercut
Mayo Scissors Supercut, 17.0 cm (6.6"), Stainless steel, Straight
Catalog # 77663-310
Supplier #:  503384
Mayo Scissors Supercut
Catalog # 77663-310
Mayo Scissors Supercut
Mayo Scissors Supercut, 17.0 cm (6.6"), Stainless steel, Straight
Price
$530.39
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Supercut mayo scissors, also called mayo dissecting scissors, are surgical scissors made popular by the mayo clinic. They have ring handles and long, straight, thick blades with blunt tips that are straight or curved. The surgical scissors vary in length from 14 cm (5.5") to 25 cm (9.8"). Longer blades give you greater reach for deeper dissection. The blades are sturdy for heavy duty work like cutting connective tissues, fascia, and sutures. Mayo scissors are often used for dissection, where the blunt tips prevent accidental damage to surrounding tissue and may be used to delicately probe. They may also be used for dressing and bandage removal.
  • Supercut scissors have one edge micro serrated, one edge honed to the sharpness of a knife
  • Blunt, thick tips
  • Ring handles
  • Straight or curved tips
  • Surgical grade stainless steel or premium German steel
As indicated by the black handle, these surgical scissors are supercut, which means that they have one blade that is micro serrated to gently grasp the tissue and one edge that is honed razor sharp for a clean cut. Supercut scissors reduce tissue trauma, offer you a more controlled cutting motion, and give you superior cutting ability.
These surgical scissors are available in surgical grade stainless steel or premium German steel. Both metals are suitable for sterilization by autoclaving or chemically. Always clean your surgical instruments promptly after use. Cleaning helps to minimize staining and preserve the life of your fine surgical instruments.
Specifications
  • Désignation:
    Mayo Scissors Supercut
  • Longueur:
    17.0 cm (6.6")
  • Matière:
    Stainless steel
  • Curvature:
    Straight