Jansen Retractor | Avantor
Jansen Retractor
11.5 cm (4.5"), German steel
Catalog # 77662-832
Fournisseur #:  501770-G
Jansen Retractor
Catalog # 77662-832
Jansen Retractor
11.5 cm (4.5"), German steel
Price
$227.52
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Jansen retractor is a commonly used to provide and maintain exposure of a surgical site. The Jansen retractor is a self-retaining retractor, meaning it can hold tissues and organs in place without someone to manually hold it.
  • 3×3 blunt prongs
  • Maximum spread of 3 cm
  • Autoclavable
  • Stainless steel or premium German steel
The frame has an adjustment screw on the top, allowing you to adjust the opening of the jaws when you position it and to lock it in place. The blades are attached to the frame and are used to retract tissues and hold them aside during the procedure.
Jansen retractors are commonly used in abdominal and pelvic surgeries, particularly for procedures that require a large incision or involve deep access to organs. They are useful for creating and maintaining a clear and unobstructed surgical field, allowing the surgeon to visualize and access the area of interest effectively. By retracting tissues and organs, the retractor helps improve visibility, facilitates the manipulation of structures, and provides space for surgical instruments to work.
Spécifications
  • Longueur:
    11.5 cm (4.5")
  • Matière:
    German steel