Iris Scissors, Ribbon Handle | Avantor
Iris Scissors, Ribbon Handle
Iris scissor, curved tip, 11 cm (4.3")
Catalog # 77662-828
Fournisseur #:  501768
Iris Scissors, Ribbon Handle
Catalog # 77662-828
Iris Scissors, Ribbon Handle
Iris scissor, curved tip, 11 cm (4.3")
Price
$146.01
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved or straight styles.
  • Large ring handles
  • Stainless steel
Spécifications
  • Désignation:
    Iris scissor, curved tip
  • Longueur:
    11 cm (4.3")