Iris Scissors, Ribbon Handle
Iris Scissors, Ribbon Handle
Iris scissor, straight tip, 11 cm (4.3")
Catalog # 77662-826
Supplier #:  501767
Iris Scissors, Ribbon Handle
Catalog # 77662-826
Iris Scissors, Ribbon Handle
Iris scissor, straight tip, 11 cm (4.3")
Price
$143.57
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved or straight styles.
  • Large ring handles
  • Stainless steel
Specifications
  • Désignation:
    Iris scissor, straight tip
  • Longueur:
    11 cm (4.3")