Absorbent Paper Points
Absorbent paper points are commonly used in laboratories, particularly in the field of dentistry and endodontics. These paper points are small, sterile, and highly absorbent.
- Made from higly absorbent paper
- Uniformly rolled to ensure correct size
- Good for small volume absorption in confined areas
These sterile paper points are tapered to target specific areas and soak up small volumes of liquid. A variety of color-coded sizes are available, including extra fine, #15 fine, medium and coarse.
: Package of 100 to 200, depending on the size of the paper points.
- Color:White
- Packaging:Pack of 200
- Type:Fine