Senn Retractor
The Senn Retractor is a hand retractor which lets to gently hold back or retract tissues for better visibility and access to the surgical site.
- Flat, thin, and curved blades
- Sharp or blunt tips
- Double ended
- Fenestrated and solid blades
- Surgical grade stainless steel which is autoclavable
This is commonly used in procedures involving skin, subcutaneous tissues, or superficial muscles. The retractor's blades can be inserted between layers of tissue and gently maneuvered to create the desired exposure. Use it for wound exposure, soft tissue dissection, or orthopedic applications.
The two ended handle has both fenestrated and solid blades which are both flat, thin, and curved to facilitate tissue retraction and maneuverability.
- Longueur:15.9 cm (6.25")
- Matière:Stainless steel
- Tip Configuration:Shart tip