Iris Scissors, Supercut
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
- Supercut-one edge micro serrated, one edge honed to the sharpness of a knife
- Sharp tips
Spécifications
- Désignation:Iris scissors
- Longueur:10 cm (4.0")
- Matière:Stainless steel
- Curvature:Straight