Reflex Clips System
Reflex Clips System
Reflex clip applier for 9 mm clips
Catalog # 77662-278
Supplier #:  500345
Reflex Clips System
Catalog # 77662-278
Reflex Clips System
Reflex clip applier for 9 mm clips
Price
$525.56
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For smal animal wound closure.
  • Clips are non stertile
  • Appliers are non sterile but can be autoclaved
Clips are made up of stainless steel material.
Packaging: Clips are packed 100 per box.
Caution: This item is designed for research only, not intended for human use.
Specifications
  • Désignation:
    Reflex clip applier for 9 mm clips