Weitlaner Retractors
Weitlaner retractor, self-retaining, 3×4 prongs, 7 cm maximum spread, sharp, 16.5 cm (6.5")
  77662-724
Weitlaner Retractors
  77662-724
Weitlaner Retractors
Weitlaner retractor, self-retaining, 3×4 prongs, 7 cm maximum spread, sharp, 16.5 cm (6.5")
$413.61
Each
Made up of stainless steel.
  • Désignation:
    Weitlaner retractor, self-retaining, 3×4 prongs, 7 cm maximum spread, sharp
  • Longueur:
    16.5 cm (6.5")