Weitlaner Retractors | Avantor
Weitlaner Retractors
Weitlaner retractor, self-retaining, 3×4 prongs, 4 cm maximum spread, sharp, German, 14 cm (5.5")
Catalog # 77662-720
Fournisseur #:  501721-G
Weitlaner Retractors
Catalog # 77662-720
Weitlaner Retractors
Weitlaner retractor, self-retaining, 3×4 prongs, 4 cm maximum spread, sharp, German, 14 cm (5.5")
Price
$428.20
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made up of stainless steel.
Spécifications
  • Désignation:
    Weitlaner retractor, self-retaining, 3×4 prongs, 4 cm maximum spread, sharp, German
  • Longueur:
    14 cm (5.5")