Metzenbaum Scissors
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue.
The tips are semi-blunt, and they are available in curved or straight styles.
- Désignation:Metzenbaum scissors, straight tips
- Longueur:17.5 cm (7")
- Matière:German steel