Operating Scissors
Operating scissors, 16 cm (6.25"), Premium German steel
  77662-418
 :  501223-G
Operating Scissors
  77662-418
Operating Scissors
Operating scissors, 16 cm (6.25"), Premium German steel
$79.00
Each
Ring-handled operating scissors are operating scissors are available with straight or curved blades commonly used in general surgery for cutting and dissecting tissues.
  • Sharp/sharp, Sharp/blunt, or Blunt/blunt tips
  • Stainless steel or premium German steel
  • Autoclavable
That can be used for a variety of procedures, including biological and medical research. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades.
They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion. Operating scissors are designed to withstand the high heat and chemical exposure of the sterilization process.
  • Désignation:
    Operating scissors
  • Longueur:
    16 cm (6.25")
  • Matière:
    Premium German steel