MICRO-ePORE™ Electrode Holders | Avantor
MICRO-ePORE™ Electrode Holders
WPI MICRO-ePORE pinpoint cell penetrator when using a Narishige injector, 1.0 mm pipette gaskets (lime green), 1.2 mm pipette gaskets (black), 1.5 mm pipette gaskets (red), 1.65 mm pipette gaskets (white), sealing gaskets, Narishige MICRO-ePORE holder, silver wire
Catalog # 77662-062
Fournisseur #:  300685
MICRO-ePORE™ Electrode Holders
Catalog # 77662-062
MICRO-ePORE™ Electrode Holders
WPI MICRO-ePORE pinpoint cell penetrator when using a Narishige injector, 1.0 mm pipette gaskets (lime green), 1.2 mm pipette gaskets (black), 1.5 mm pipette gaskets (red), 1.65 mm pipette gaskets (white), sealing gaskets, Narishige MICRO-ePORE holder, silver wire
Price
$877.53
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These electrode holders are designed exclusively for use the with MICRO-ePORE™ system, which includes the silver wire electrode holders. The platinum wire electrode holders are an optional accessory.
Platinum wire electrode holders are important when you are working with proteins. The silver chloride wire is perfect for work with RNA. However, the silver chloride wire tends to denature proteins, reducing the efficiency of the microinjection solution.
Applications for silver wire electrode holders: Silver wire electrodes are suitable for most applications
Applications for platinum wire electrode holders: Platinum electrodes offers more active insertion of the genetic material capability, and there is a better chance of changing the genome
Spécifications
  • Désignation:
    Narishige MICRO-ePORE holder, silver wire
  • For Use With:
    WPI MICRO-ePORE pinpoint cell penetrator when using a Narishige injector
  • Includes:
    1.0 mm pipette gaskets (lime green), 1.2 mm pipette gaskets (black), 1.5 mm pipette gaskets (red), 1.65 mm pipette gaskets (white), sealing gaskets