Dissecting Scissors | Avantor
Dissecting Scissors
Dissecting scissors, 12.5 cm (4"), curved, Stainless steel
Catalog # 77661-958
Fournisseur #:  15923
Dissecting Scissors
Catalog # 77661-958
Dissecting Scissors
Dissecting scissors, 12.5 cm (4"), curved, Stainless steel
Price
$99.78
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ring-handled dissecting scissors are surgical scissors commonly used to cut and separate delicate tissues. These scissors are 10 or 12.5 cm long. These dissecting scissors are available with straight or curved blades. These dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors.
  • Multi-purpose use
  • Ideal for cutting soft tissue or cartilage
  • Straight or curved
  • Long thin blades make them lighter than operating scissors
  • Surgical grade stainless steel or premium German steel
They are designed for cutting and probing in tight spaces. Their tips are pointed, where operating scissors are generally blunt. These are available in surgical grade stainless steel or premium German steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
Spécifications
  • Désignation:
    Dissecting scissors, 12.5 cm (4"), curved
  • Matière:
    Stainless steel