BNC Right Angles
Standard BNC cable, connector A: BNC (male), connector B: BNC (male), cable length: 15 cm (6")
  77662-254
BNC Right Angles
  77662-254
BNC Right Angles
Standard BNC cable, connector A: BNC (male), connector B: BNC (male), cable length: 15 cm (6")
$26.80
Each
  • Connector A: BNC (male)
  • Connector B: BNC (female)
  • Cable: none
  • Désignation:
    Standard BNC cable, connector A: BNC (male), connector B: BNC (male), cable length: 15 cm (6")