BNC Right Angles | Avantor
BNC Right Angles
BNC right angle adapter, connector A: BNC (male), connector B: BNC (female)
Catalog # 77662-252
Fournisseur #:  500256
BNC Right Angles
Catalog # 77662-252
BNC Right Angles
BNC right angle adapter, connector A: BNC (male), connector B: BNC (female)
Price
$26.80
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Connector A: BNC (male)
  • Connector B: BNC (female)
  • Cable: none
Spécifications
  • Désignation:
    BNC right angle adapter, connector A: BNC (male), connector B: BNC (female)