Iris Scissors, Supercut
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
- Supercut-one edge micro serrated, one edge honed to the sharpness of a knife
- Sharp tips
- Désignation:Iris scissors
- Longueur:12.5 cm (5.0")
- Matière:German steel
- Curvature:Curved