Tenotomy Scissors | Avantor
Tenotomy Scissors
Supercut scissor, curved, 12.5 cm, Stainless steel
Catalog # 77661-862
Fournisseur #:  14221
Tenotomy Scissors
Catalog # 77661-862
Tenotomy Scissors
Supercut scissor, curved, 12.5 cm, Stainless steel
Price
$226.29
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tenotomy scissors are used for delicate procedures. With the long handle and thin tapered blades, the tenotomy scissors are able to cut tissues in hard to reach, limited spaces.
  • One serrated blade, one honed razor sharp
  • Sharp tips
  • Surgical grade stainless steel
These sharp, straight scissors make small, precision cuts, and they are perfect for delicate operations. The longer handles help to control the precision of these tenotomy scissors in restricted spaces or with delicate membranes or tissues, like brain tissue or ophthalmic surgeries.
These scissors are designed for probing and cutting in tight spots. A precision pair of scissors like the tenotomy scissors limits the damage to surrounding tissues, which is critical in neurosurgery and other applications.
Spécifications
  • Désignation:
    Supercut scissor, curved
  • Longueur:
    12.5 cm
  • Matière:
    Stainless steel