Reflex Clips System
Reflex clip applier for 7 mm clips
  77662-274
Reflex Clips System
  77662-274
Reflex Clips System
Reflex clip applier for 7 mm clips
$525.56
Each
For smal animal wound closure.
  • Clips are non stertile
  • Appliers are non sterile but can be autoclaved
Clips are made up of stainless steel material.
 : Clips are packed 100 per box.
 : This item is designed for research only, not intended for human use.
  • Désignation:
    Reflex clip applier for 7 mm clips