BNC Right Angles | Avantor
BNC Right Angles
Standard BNC cable, connector A: BNC (male), connector B: BNC (male), cable length: 46 cm (18")
Catalog # 77662-256
Fournisseur #:  500259
BNC Right Angles
Catalog # 77662-256
BNC Right Angles
Standard BNC cable, connector A: BNC (male), connector B: BNC (male), cable length: 46 cm (18")
Price
$26.80
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Connector A: BNC (male)
  • Connector B: BNC (female)
  • Cable: none
Spécifications
  • Désignation:
    Standard BNC cable, connector A: BNC (male), connector B: BNC (male), cable length: 46 cm (18")