Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

GBS Detect/Strep Reagents

GBS Detect™, for non-hemolytic group B Strep, 15×100 mm plate, 100 plates/box
Catalog # CA77623-656
 :  A300BX
GBS Detect/Strep Reagents
Catalog # CA77623-656
Supplier:  
CAS Number:  
GBS Detect/Strep Reagents
GBS Detect/Strep Reagents

GBS Detect™, for non-hemolytic group B Strep, 15×100 mm plate, 100 plates/box

CAS Number:  
Discontinued
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

GBS Detect is used for the isolation and differentiation by enhanced hemolysis of gamma hemolytic Group B Streptococcus.

  • Grouping reagents for the rapid latex agglutination test
  • Lancefield grouping includes A,B,C,D,F,G latex, controls, extraction reagents, cards, and mixing sticks
  • StrepQuick™ to aid in the identification of gram positive cocci

GBS Detect™, for non-hemolytic Group B Strep. Granada Medium for hemolytic Group B Strep. Grouping Reagents A, B, C, and G, for the rapid latex agglutination test. PYR Test Kit with chromogenic developer on paper disks. Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), especially for throat cultures. Selective Strep Agar (COBA Medium), for the selective isolation of all species of Strep. ci. StrepQuick™ performs a PYR, LAP, and Esculin test to aid in the identification of gram positive coc.

Spécifications

Spécifications

  • Désignation:
    GBS Detect™, for non-hemolytic group B Strep
  • Taille:
    100 plates/box
  • Format:
    15×100 mm plate

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.