Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
1M Sodium Phosphate Buffer | Avantor

1M Sodium Phosphate Buffer

1M sodium phosphate buffer, 1000 ml, Plastic bottle, 7.2
Catalog # CA77442-714
Supplier:   Teknova
Fournisseur #:  P3072
1M Sodium Phosphate Buffer
Catalog # CA77442-714
Supplier:  Teknova
CAS Number:  
1M Sodium Phosphate Buffer

1M sodium phosphate buffer, 1000 ml, Plastic bottle, 7.2

CAS Number:  
Price
$82.30
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

312 mM sodium phosphate monobasic, 688 mM sodium phosphate dibasic.

Spécifications

Spécifications

  • Type de conditionnement:
    Plastic bottle
  • Désignation:
    1M sodium phosphate buffer
  • Taille:
    1000 ml
  • pH:
    7.2
  • Cat. No.:
    CA77442-714
  • Sterility:
    Sterile

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.