VWR® pH Electrodes | Avantor
VWR® pH Electrodes
pH composite electrode (HF acid titrate), Polypropylene, S7 [S7-BNC(Q9) cable], 3 M KCl, 0...60 °C, 2 - 11
Catalog # 77618-906
Supplier:   VWR International
VWR® pH Electrodes
Catalog # 77618-906
Supplier:  VWR International
VWR® pH Electrodes
VWR® pH Electrodes
pH composite electrode (HF acid titrate), Polypropylene, S7 [S7-BNC(Q9) cable], 3 M KCl, 0...60 °C, 2 - 11
Price
$505.43
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
The VWR® collection pH electrodes provide a wide-array of sensor options for any application requirement.
  • Specialized sensor configurations for various applications including wastewater, soil, food and beverage, and micro-container measurements
  • BNC connector (BNC(Q9) or S7)
  • Variety of body materials including glass, lead-free glass, polycarbonate, and epoxy
  • Gel or liquid electrolyte fill solution
  • Includes unique serial number
  • Easily connect to VWR(R) pH meters
For additional specifications and application specific recommendations, please reference 'VWR® pH Electrode Selection Guide' located in the Documents tab below.
Spécifications
  • Plage de température:
    0...60 °C
  • Type:
    pH composite electrode (HF acid titrate)
  • Matière:
    Polypropylene
  • pH Range:
    2 - 11
  • Fill Solution:
    3 M KCl
  • Connecteur:
    S7 [S7-BNC(Q9) cable]
Accessories
Informations sur le produit
  • Catalog #
    Material
    For Use With
    Availability
    Price
  • VWR® ATC probe
    VWR® ATC probe
    77618-918
    316 stainless steel
    VWR® 500-, 550-, 700-, 750-, 900, 950-series meters
    Log in pour la disponibilité
    Each - $163.22