VWR® pH Electrodes
pH composite electrode (Ultrapure water), Glass, S7 [S7-BNC(Q9) cable], 3 M KCl, 0...80 °C, 0 - 11
Catalog # 77618-900
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® collection pH electrodes provide a wide-array of sensor options for any application requirement.
Specialized sensor configurations for various applications including wastewater, soil, food and beverage, and micro-container measurements
- BNC connector (BNC(Q9) or S7)
- Variety of body materials including glass, lead-free glass, polycarbonate, and epoxy
- Gel or liquid electrolyte fill solution
- Includes unique serial number
- Easily connect to VWR(R) pH meters
For additional specifications and application specific recommendations, please reference 'VWR® pH Electrode Selection Guide' located in the Documents tab below.
Spécifications
- Plage de température:0...80 °C
- Type:pH composite electrode (Ultrapure water)
- Matière:Glass
- pH Range:0 - 11
- Fill Solution:3 M KCl
- Connecteur:S7 [S7-BNC(Q9) cable]
Accessories
Informations sur le produit
- Catalog #MaterialFor Use WithAvailabilityPrice
- VWR® ATC probeVWR® ATC probe77618-918316 stainless steelVWR® 500-, 550-, 700-, 750-, 900, 950-series metersLog in pour la disponibilitéEach - $163.22