Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

VWR® pH Electrodes

pH composite electrode (Low maintenance), Polycarbonate, BNC(Q9), Unfillable, 0...80 °C, 0 - 14
Catalog # 77618-894
Supplier:   VWR International
VWR® pH Electrodes
Catalog # 77618-894
Supplier:  VWR International
VWR® pH Electrodes
VWR® pH Electrodes

pH composite electrode (Low maintenance), Polycarbonate, BNC(Q9), Unfillable, 0...80 °C, 0 - 14

Price
$515.40
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

The VWR® collection pH electrodes provide a wide-array of sensor options for any application requirement.

  • Specialized sensor configurations for various applications including wastewater, soil, food and beverage, and micro-container measurements

  • BNC connector (BNC(Q9) or S7)
  • Variety of body materials including glass, lead-free glass, polycarbonate, and epoxy
  • Gel or liquid electrolyte fill solution
  • Includes unique serial number
  • Easily connect to VWR(R) pH meters

For additional specifications and application specific recommendations, please reference 'VWR® pH Electrode Selection Guide' located in the Documents tab below.

Spécifications
  • Plage de température:
    0...80 °C
  • Type:
    pH composite electrode (Low maintenance)
  • Matière:
    Polycarbonate
  • pH Range:
    0 - 14
  • Fill Solution:
    Unfillable
  • Connecteur:
    BNC(Q9)
Accessories

Informations sur le produit

  • Catalog #
    Material
    For Use With
    Availability
    Price
  • VWR® ATC probe
    VWR® ATC probe
    77618-918
    316 stainless steel
    VWR® 500-, 550-, 700-, 750-, 900, 950-series meters
    Log in pour la disponibilité
    Each - $163.22