Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® pH Electrodes | Avantor

VWR® pH Electrodes

3-in-1 pH composite electrode (built-in ATC), Polycarbonate, BNC(Q9) and 4-pin aviation, 3 M KCl, 5...60 °C, 0 - 14
Catalog # 77618-868
Supplier:   VWR International
VWR® pH Electrodes
Catalog # 77618-868
Supplier:  VWR International
VWR® pH Electrodes
VWR® pH Electrodes

3-in-1 pH composite electrode (built-in ATC), Polycarbonate, BNC(Q9) and 4-pin aviation, 3 M KCl, 5...60 °C, 0 - 14

Price
$254.50
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

The VWR® collection pH electrodes provide a wide-array of sensor options for any application requirement.

  • Specialized sensor configurations for various applications including wastewater, soil, food and beverage, and micro-container measurements

  • BNC connector (BNC(Q9) or S7)
  • Variety of body materials including glass, lead-free glass, polycarbonate, and epoxy
  • Gel or liquid electrolyte fill solution
  • Includes unique serial number
  • Easily connect to VWR(R) pH meters

For additional specifications and application specific recommendations, please reference 'VWR® pH Electrode Selection Guide' located in the Documents tab below.

Spécifications
  • Plage de température:
    5...60 °C
  • Type:
    3-in-1 pH composite electrode (built-in ATC)
  • Matière:
    Polycarbonate
  • pH Range:
    0 - 14
  • Fill Solution:
    3 M KCl
  • Connecteur:
    BNC(Q9) and 4-pin aviation
Accessories

Informations sur le produit

  • Catalog #
    Material
    For Use With
    Availability
    Price
  • VWR® ATC probe
    VWR® ATC probe
    77618-918
    316 stainless steel
    VWR® 500-, 550-, 700-, 750-, 900, 950-series meters
    Log in pour la disponibilité
    Each - $163.22

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.