VWR® Ion Selectrive Electrodes
VWR® ISE15 ion selective electrode, combination, BNC (Q9), Plastic, (0.01 - 20) ppm (Cl₂), 0…60 °C, Residual chlorine
Catalog # 77618-860
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
Spécifications
- Plage de température:0…60 °C
- Désignation:VWR® ISE15 ion selective electrode, combination
- Measurement Range:(0.01 - 20) ppm (Cl₂)
- Body Material:Plastic
- Ion:Residual chlorine
- Connecteur:BNC (Q9)