VWR® Ion Selectrive Electrodes
VWR® Ion Selectrive Electrodes
VWR® ISE13 ion selective electrode, combination, BNC (Q9), Plastic, 10⁻¹ - 10⁻⁵ M, 0…40 °C, Perchlorate (ClO₄ )
Catalog # 77618-856
Supplier:   VWR International
VWR® Ion Selectrive Electrodes
Catalog # 77618-856
Supplier:  VWR International
VWR® Ion Selectrive Electrodes
VWR® Ion Selectrive Electrodes
VWR® ISE13 ion selective electrode, combination, BNC (Q9), Plastic, 10⁻¹ - 10⁻⁵ M, 0…40 °C, Perchlorate (ClO₄ )
Price
$1,072.74
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
  • ISE probe and reference electrode combined
  • BNC connection
Includes ISE reference filler solution.
Specifications
  • Plage de température:
    0…40 °C
  • Désignation:
    VWR® ISE13 ion selective electrode, combination
  • Measurement Range:
    10⁻¹ - 10⁻⁵ M
  • Body Material:
    Plastic
  • Ion:
    Perchlorate (ClO₄ )
  • Connecteur:
    BNC (Q9)