VWR® Ion Selectrive Electrodes
VWR® ISE13 ion selective electrode, combination, BNC (Q9), Plastic, 10⁻¹ - 10⁻⁵ M, 0…40 °C, Perchlorate (ClO₄ )
Catalog # 77618-856
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
Specifications
- Plage de température:0…40 °C
- Désignation:VWR® ISE13 ion selective electrode, combination
- Measurement Range:10⁻¹ - 10⁻⁵ M
- Body Material:Plastic
- Ion:Perchlorate (ClO₄ )
- Connecteur:BNC (Q9)