VWR® Ion Selectrive Electrodes
VWR® ISE12 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁶ - 0.1 M, 0…40 °C, Fluoroborate (BF₄)
Catalog # 77618-854
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
Spécifications
- Plage de température:0…40 °C
- Désignation:VWR® ISE12 ion selective electrode, combination
- Measurement Range:1.0×10⁻⁶ - 0.1 M
- Body Material:Plastic
- Ion:Fluoroborate (BF₄)
- Connecteur:BNC (Q9)