VWR® Ion Selectrive Electrodes
VWR® Ion Selectrive Electrodes
VWR® ISE10 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁵ - 1.0 M, 0…40 °C, Nitrate (NO³⁻)
  77618-850
VWR® Ion Selectrive Electrodes
  77618-850
VWR® Ion Selectrive Electrodes
VWR® Ion Selectrive Electrodes
VWR® ISE10 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁵ - 1.0 M, 0…40 °C, Nitrate (NO³⁻)
$1,072.74
Each
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
  • ISE probe and reference electrode combined
  • BNC connection
Includes ISE reference filler solution.
  • Plage de température:
    0…40 °C
  • Désignation:
    VWR® ISE10 ion selective electrode, combination
  • Measurement Range:
    1.0×10⁻⁵ - 1.0 M
  • Body Material:
    Plastic
  • Ion:
    Nitrate (NO³⁻)
  • Connecteur:
    BNC (Q9)