VWR® Ion Selectrive Electrodes
VWR® ISE10 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁵ - 1.0 M, 0…40 °C, Nitrate (NO³⁻)
77618-850
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
- Plage de température:0…40 °C
- Désignation:VWR® ISE10 ion selective electrode, combination
- Measurement Range:1.0×10⁻⁵ - 1.0 M
- Body Material:Plastic
- Ion:Nitrate (NO³⁻)
- Connecteur:BNC (Q9)