VWR® Ion Selectrive Electrodes
VWR® ISE09 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁶ - 1.0 M, 0…40 °C, Ammonium (NH₄)
Catalog # 77618-848
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
Spécifications
- Plage de température:0…40 °C
- Désignation:VWR® ISE09 ion selective electrode, combination
- Measurement Range:1.0×10⁻⁶ - 1.0 M
- Body Material:Plastic
- Ion:Ammonium (NH₄)
- Connecteur:BNC (Q9)