VWR® Ion Selectrive Electrodes | Avantor
VWR® Ion Selectrive Electrodes
VWR® ISE09 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁶ - 1.0 M, 0…40 °C, Ammonium (NH₄)
Catalog # 77618-848
Supplier:   VWR International
VWR® Ion Selectrive Electrodes
Catalog # 77618-848
Supplier:  VWR International
VWR® Ion Selectrive Electrodes
VWR® Ion Selectrive Electrodes
VWR® ISE09 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁶ - 1.0 M, 0…40 °C, Ammonium (NH₄)
Price
$1,072.74
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
  • ISE probe and reference electrode combined
  • BNC connection
Includes ISE reference filler solution.
Spécifications
  • Plage de température:
    0…40 °C
  • Désignation:
    VWR® ISE09 ion selective electrode, combination
  • Measurement Range:
    1.0×10⁻⁶ - 1.0 M
  • Body Material:
    Plastic
  • Ion:
    Ammonium (NH₄)
  • Connecteur:
    BNC (Q9)