VWR® Ion Selectrive Electrodes | Avantor
VWR® Ion Selectrive Electrodes
VWR® ISE07 ion selective electrode, combination, S7, Glass, 1.0×10⁻⁶ - saturation, 0…80 °C, Sodium (Na⁺)
Catalog # 77618-844
Supplier:   VWR International
VWR® Ion Selectrive Electrodes
Catalog # 77618-844
Supplier:  VWR International
VWR® Ion Selectrive Electrodes
VWR® Ion Selectrive Electrodes
VWR® ISE07 ion selective electrode, combination, S7, Glass, 1.0×10⁻⁶ - saturation, 0…80 °C, Sodium (Na⁺)
Price
$934.53
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
  • ISE probe and reference electrode combined
  • BNC connection
Includes ISE reference filler solution.
Spécifications
  • Plage de température:
    0…80 °C
  • Désignation:
    VWR® ISE07 ion selective electrode, combination
  • Measurement Range:
    1.0×10⁻⁶ - saturation
  • Body Material:
    Glass
  • Ion:
    Sodium (Na⁺)
  • Connecteur:
    S7