VWR® Ion Selectrive Electrodes
VWR® ISE07 ion selective electrode, combination, S7, Glass, 1.0×10⁻⁶ - saturation, 0…80 °C, Sodium (Na⁺)
Catalog # 77618-844
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
Spécifications
- Plage de température:0…80 °C
- Désignation:VWR® ISE07 ion selective electrode, combination
- Measurement Range:1.0×10⁻⁶ - saturation
- Body Material:Glass
- Ion:Sodium (Na⁺)
- Connecteur:S7