VWR® Ion Selectrive Electrodes
VWR® ISE06 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁶ - 0.1 M, 0…80 °C, Lead (Pb²⁺)
77618-842
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
- Plage de température:0…80 °C
- Désignation:VWR® ISE06 ion selective electrode, combination
- Measurement Range:1.0×10⁻⁶ - 0.1 M
- Body Material:Plastic
- Ion:Lead (Pb²⁺)
- Connecteur:BNC (Q9)