VWR® Ion Selectrive Electrodes
VWR® ISE02 ion selective electrode, combination, BNC (Q9), Plastic, 5.0×10⁻⁵ - 1.0 M, 0…80 °C, Chloride (Cl⁻)
Catalog # 77618-834
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
Spécifications
- Plage de température:0…80 °C
- Désignation:VWR® ISE02 ion selective electrode, combination
- Measurement Range:5.0×10⁻⁵ - 1.0 M
- Body Material:Plastic
- Ion:Chloride (Cl⁻)
- Connecteur:BNC (Q9)