Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
phi29-XT WGA Kit | Avantor

phi29-XT WGA Kit

phi29-XT WGA kit, 25
Catalog # 77591-838
Fournisseur #:  E1604S
phi29-XT WGA Kit
Catalog # 77591-838
CAS Number:  
phi29-XT WGA Kit
phi29-XT WGA Kit

phi29-XT WGA kit, 25

CAS Number:  
Price
$346.03
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

The phi29-XT WGA Kit has been optimized for sensitive, fast, and robust whole genome amplification (WGA), an isothermal method for amplification of an entire genome, starting from femtogram quantities of DNA and resulting in microgram quantities of amplified products.

  • High sensitivity: Amplify from as little as 10 fg of input DNA
  • Powered by phi29-XT DNA Polymerase, an engineered polymerase
  • Flexible protocol allows for different types of input material
  • Simple workflow produces high yield in a short reaction time (less than 2 hours)
  • Whole genome representation and uniform GC coverage for human and microbial DNA

The kit features phi29-XT DNA polymerase, an engineered polymerase with improved thermostability, sensitivity, and generates higher yield in a shorter reaction time than wild-type phi29 DNA Polymerase. Also included are exonuclease-resistant random primers (containing phosphorothioate bonds) to universally amplify any DNA sequence, neutralization buffer, and dNTPs.

Input material can be purified genomic DNA, cells, or microbiomes. WGA enables several downstream applications, including single cell whole genome sequencing (WGS), genomic DNA enrichment, and environmental DNA (eDNA) enrichment and sequencing.

Spécifications

Spécifications

  • Désignation:
    phi29-XT WGA kit
  • No. of Reactions:
    25

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.