Dynamics Track System
Product Details & Documents
This cost-effective dynamics system is a versatile tool for studying linear motion and collisions.
Available with and without photogates
- Comes with a large assortment of accessories to facilitate many different types of experiments
A dynamic cart and track system is an advanced physics lab tool used to study motion, forces, momentum, and energy with high-precision timing measurements. Measure velocity, acceleration, and collision events with high accuracy. Newton’s Laws experiments - Demonstrate inertia, force, and action-reaction principles. Momentum and Energy Conservation - Study elastic and inelastic collisions with real-time data collection. Kinematic studies - Analyze velocity, acceleration, and free-fall motion with detailed measurements. Friction and Inclined Plane investigations - Examine how forces and motion change with different surfaces or slopes.
Spécifications
- Désignation:Dynamics track system with photogates
- Longueur:120 cm