Dynamics Track System | Avantor
Dynamics Track System
Dynamics track system with photogates, 120 cm
Catalog # 470356-990
Supplier:   GSC International
Fournisseur #:  DCTS-2
Dynamics Track System
Catalog # 470356-990
Supplier:  GSC International
Dynamics Track System
Dynamics Track System
Dynamics track system with photogates, 120 cm
Price
$971.70
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
This cost-effective dynamics system is a versatile tool for studying linear motion and collisions.
  • Available with and without photogates
  • Comes with a large assortment of accessories to facilitate many different types of experiments
A dynamic cart and track system is an advanced physics lab tool used to study motion, forces, momentum, and energy with high-precision timing measurements. Measure velocity, acceleration, and collision events with high accuracy. Newton’s Laws experiments - Demonstrate inertia, force, and action-reaction principles. Momentum and Energy Conservation - Study elastic and inelastic collisions with real-time data collection. Kinematic studies - Analyze velocity, acceleration, and free-fall motion with detailed measurements. Friction and Inclined Plane investigations - Examine how forces and motion change with different surfaces or slopes.
Spécifications
  • Désignation:
    Dynamics track system with photogates
  • Longueur:
    120 cm