Dynamics Track System | Avantor
Dynamics Track System
Dynamics track system, 120 cm
Catalog # 470357-026
Supplier:   GSC International
Fournisseur #:  DCTS-1
Dynamics Track System
Catalog # 470357-026
Supplier:  GSC International
Dynamics Track System
Dynamics Track System
Dynamics track system, 120 cm
Price
$711.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
This cost-effective dynamics system is a versatile tool for studying linear motion and collisions.
  • Available with and without photogates
  • Comes with a large assortment of accessories to facilitate many different types of experiments
A dynamic cart and track system is an advanced physics lab tool used to study motion, forces, momentum, and energy with high-precision timing measurements. Measure velocity, acceleration, and collision events with high accuracy. Newton’s Laws experiments - Demonstrate inertia, force, and action-reaction principles. Momentum and Energy Conservation - Study elastic and inelastic collisions with real-time data collection. Kinematic studies - Analyze velocity, acceleration, and free-fall motion with detailed measurements. Friction and Inclined Plane investigations - Examine how forces and motion change with different surfaces or slopes.
Spécifications
  • Désignation:
    Dynamics track system
  • Longueur:
    120 cm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.