PathFlow PiSmart Laser Cassettes, Threaded
Product Details & Documents
Pathflow PiSmart laser cassettes are a two piece design with a rear hinge and front tab closure. Threaded cassettes come in stacks of 40 cassettes, and 50 stacks per case of 2000.
These threaded cassettes can be easily and quickly loaded in the PiSmart Laser Printers. These cassettes are a validated consumable for the PiSmart laser cassette printer.
Spécifications
- Color:Pink
- Désignation:PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded