PathFlow PiSmart Laser Cassettes, Threaded | Avantor
PathFlow PiSmart Laser Cassettes, Threaded
PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded, Orange
Catalog # 77575-554
Supplier:   StatLab
Fournisseur #:  CAS-LPI100BX-OR
PathFlow PiSmart Laser Cassettes, Threaded
Catalog # 77575-554
Supplier:  StatLab
PathFlow PiSmart Laser Cassettes, Threaded
PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded, Orange
Price
$1,419.00
Log in pour la disponibilité
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pathflow PiSmart laser cassettes are a two piece design with a rear hinge and front tab closure. Threaded cassettes come in stacks of 40 cassettes, and 50 stacks per case of 2000.
These threaded cassettes can be easily and quickly loaded in the PiSmart Laser Printers. These cassettes are a validated consumable for the PiSmart laser cassette printer.
Spécifications
  • Color:
    Orange
  • Désignation:
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded