Cleanroom Labels, Core2Write
Core2Write label, sterile, irradiated, 101,6×76,2 mm, 6 labels/sheet, 500 sheets/case
Catalog # 76627-995
Supplier: Veltek Associates
Fournisseur #: C2WR-4X3-SHEET-02
Jump to:
Product Details & Documents
Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
Spécifications
- Désignation:Core2Write label, sterile, irradiated
- Dimensions:101,6×76,2 mm
- Packaging:6 labels/sheet, 500 sheets/case