Cleanroom Labels, Core2Write
Core2Write label, sterile, irradiated, 76,2×25,4 mm, 18 labels/sheet, 500 sheets/case
Catalog # 76627-991
Supplier: Veltek Associates
: C2WR-3X1-SHEET-02
Jump to:
Product Details & Documents
Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
Spécifications
- Désignation:Core2Write label, sterile, irradiated
- Dimensions:76,2×25,4 mm
- Packaging:18 labels/sheet, 500 sheets/case