Cleanroom Labels, Core2Write | Avantor
Cleanroom Labels, Core2Write
Core2Write label, non-sterile, 50,8×25,4 mm, 10 labels/sheet/500 sheets/case
Catalog # 76627-989
Supplier:   Veltek Associates
Fournisseur #:  C2WR-2X1-SHEET-01
Cleanroom Labels, Core2Write
Catalog # 76627-989
Supplier:  Veltek Associates
Cleanroom Labels, Core2Write
Core2Write label, non-sterile, 50,8×25,4 mm, 10 labels/sheet/500 sheets/case
Price
$5,480.00
Log in pour la disponibilité
Case of 5,000
Product Details & Documents
Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
Spécifications
  • Désignation:
    Core2Write label, non-sterile
  • Dimensions:
    50,8×25,4 mm
  • Packaging:
    10 labels/sheet/500 sheets/case