Logic® Washable Prefilter Kits | Avantor
Logic® Washable Prefilter Kits
Washable prefilter kit, 92.1×73.7×15.7 cm (36.25W×29D×6.2"H)
Catalog # 77574-562
Supplier:   Labconco
Fournisseur #:  3288600
Logic® Washable Prefilter Kits
Catalog # 77574-562
Supplier:  Labconco
Logic® Washable Prefilter Kits
Logic® Washable Prefilter Kits
Washable prefilter kit, 92.1×73.7×15.7 cm (36.25W×29D×6.2"H)
Price
$356.00
Log in pour la disponibilité
Each
Product Details & Documents
Washable prefilter kit for Logic biosafety cabinets.
  • Washable metal screen pre-filter
  • Installation hardware included
  • Compatible with Legacy Purifier Logic
  • For use with A2 and B2 biosafety cabinets
Washable Prefilters clean incoming air of dirt, dust and particles extending the life of the HEPA filter and blower motors. Each kit consists of a washable/reusable, metal screen-type prefilter and installation hardware. Legacy Purifier Logic compatible. The washable prefilter kits are designed for A2 and B2 biosafety cabinets only. Installation is required.
Spécifications
  • Désignation:
    Washable prefilter kit
  • Dimensions:
    92.1×73.7×15.7 cm (36.25W×29D×6.2"H)