Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Methyltransferase Assay Buffer IV

Methyltransferase Assay Buffer IV

Buffer for enzyme assay, 20 ml
Catalog # 77559-412
Supplier:   Sino Biological
Supplier #:  M04-09-20
Methyltransferase Assay Buffer IV
Catalog # 77559-412
Supplier:  Sino Biological
CAS Number:  
Methyltransferase Assay Buffer IV

Buffer for enzyme assay, 20 ml

CAS Number:  
Price
$171.04
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Stock solution of the buffer used for assaying protein methyltransferases.

Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.

Specifications

Specifications

  • Désignation:
    Buffer for enzyme assay
  • Taille:
    20 ml
  • Cat. No.:
    77559-412
  • Formulation:
    Frozen solution of buffer containing: 100 mM Tris-HCl, pH 8.0, 250 mM NaCl, 5 mM EDTA, 15 mM MgCl₂, 0.5 mg/ml BSA.
  • Storage temperature:
    –20 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.