Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
eQo 1-Step ToughMix® | Avantor

eQo 1-Step ToughMix®

100, 20 µl
Catalog # CA77552-856
Supplier:   Quantabio
Fournisseur #:  95301-100
eQo 1-Step ToughMix®
Catalog # CA77552-856
Supplier:  Quantabio
CAS Number:  
eQo 1-Step ToughMix®
eQo 1-Step ToughMix®

100, 20 µl

CAS Number:  
Price
$406.09
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

No compromises. Unparalleled performance meets sustainability.

  • Unrivaled sensitivity - detection down to two copies of RNA
  • Sustainable and earth friendly: Lyophilized eQo bead format eliminates dry ice, CO₂ consumptions and reduces shipping costs
  • Novel reverse transcriptase: Convenient, ambient reaction setup, overcomes rigid RNA secondary structure
  • High stability: Up to one year at room temperature, up to one year rehydrated at −20 °C
  • Tough Tested: Broad and significantly enhanced tolerance to PCR inhibitors

eQo 1-step ToughMix is a lyophilized reagent system for reverse transcription quantitative PCR (RT-qPCR) of RNA templates using hybridization probe detection chemistries such as TaqMan® 5’-hydrolysis probes. It is supplied with a proprietary rehydration buffer that when combined with the lyophilized 'eQo beads' produces a stabilized 4X concentrated master mix.

The kit includes thermolabile UDG for amplicon carryover elimination, an enhanced warm-start reverse transcriptase (RT) and all other required components for 1-step RT-qPCR except RNA template and primer/probe(s). The reaction chemistry has been optimized for inhibitor tolerance and delivers exceptional performance in either single-plex or highly demanding multiplex 1 step RT-qPCR formats.

Cycler compatibility: BioRad CFX, Quantabio Q, Roche LightCycler 480

Documents

Spécifications

Spécifications

  • No. of Reactions:
    100
  • Reaction Volume:
    20 µl
  • Multiplexing:
    Yes
  • Cat. No.:
    CA77552-856
  • Inhibitor-rich sample:
    Yes
  • One step:
    Yes
  • Application:
    RT-qPCR probe

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.