Solution d'iode de Gram
Solution d'iode de Gram
Plastic bottle, 5×250 ml
  77540-244
Supplier #:  786-2093
Solution d'iode de Gram
  77540-244
 :  12298-68-9
Solution d'iode de Gram
Plastic bottle, 5×250 ml
 :  12298-68-9
$161.24
Kit of 1
Gram Iodine Solution acts as a mordant .
  • An aqueous solution of Iodine
It is used in Gram staining to differentiate gram-positive from gram-negative bacteria
 :  Lugol's iodine, Iodine and potassium iodide, Potassium triiodide
  •  :12298-68-9
  •  :I₃K
  •  :MFCD00134128
  •  :419.812 g/mol
  • Type de conditionnement:
    Plastic bottle
  • Taille:
    5×250 ml