Triangles
Triangles, iron wire, 5.1 cm (2")
21750-095
: H-23860
These triangles are made up of iron wire (16 gauge) galvanized with plain clay pipe stems.
Smooth pipe stem triangles protect glassware on hotplates and over burner flames.
- Désignation:Triangles, iron wire, 5.1 cm (2")