Digital Force Meter
Digital Force Meter
Digital force meter, 0.1 - 20 N, single
Catalog # 470356-480
Supplier:   GSC International
Supplier #:  N-00001
Digital Force Meter
Catalog # 470356-480
Supplier:  GSC International
Digital Force Meter
Digital Force Meter
Digital force meter, 0.1 - 20 N, single
Price
$123.25
Log in pour la disponibilité
Each
Product Details & Documents
Ideal for addition and resolution of forces, Newton's laws, and statics experiments.
  • Wide measureable range and decimal resolution makes this instrument a versatile alternative to traditional spring scales
With a wide range of 2, 5, 10 and 20 Newtons, this force meter replaces four types of spring balances. With the hold function, you can retain the measured value for added convenience. With the one-touch zero-calibration function, you can measure in any direction, 360 degrees, for complete accuracy. Compact and convenient, this digital force meter measures 157 mm in length, 45 mm in width, and 24 mm in height, making it easy to use and store.
Spécifications
  • Désignation:
    Digital force meter, 0.1 - 20 N, single