QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
QuEChERS dSPE kit, TFDA method, includes animal origin, 15 ml
Catalog # AG5982-6665
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Fournisseur #: 5982-6665
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
Catalog # AG5982-6665
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CAS Number:
Jump to:
Product Details & Documents
QuEChERS dispersive solid phase extraction (SPE) kits supplied in 15 ml centrifuge tubes accommodate the aliquot volumes specified by current AOAC and EN methodologies.
- Non hazardous
The sorbent extracts the sample matrix, while MgSO₄ helps remove excess water and improve analyte partitioning.
Spécifications
Spécifications
- Capacité:15 ml
- Désignation:QuEChERS dSPE kit, TFDA method, includes animal origin